Anti-PLIN2 Mouse Monoclonal Antibody [clone: 2C5A3]
Catalog # NOVUNBP2-23486SS
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:perilipin 2
- Antigen symbol:PLIN2
- Clonality:Monoclonal
- Clone:2C5A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:123
- Antigen synonyms:ADFP|ADRP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:25 μL
- Pk:25 µl
Specifications
About this item
Type: Primary
Antigen: PLIN2 (perilipin 2)
Clonality: monoclonal
Clone: 2C5A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human