Anti-APP Mouse Monoclonal Antibody (Biotin) [clone: MOAB-2]
Catalog # NOVUNBP2-13075B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Amyloid beta (A4) Precursor Protein
- Antigen symbol:APP
- Clonality:Monoclonal
- Clone:MOAB-2
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:351
- Antigen synonyms:CTFgamma|CVAP|ABETA|APPI|AD1|PN-II|ABPP|AAA|PN2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: monoclonal
Clone: MOAB-2
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat