Anti-CHD7 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The CHD7 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CHD7. This antibody reacts with human, mouse. The CHD7 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:250
Type: Primary
Antigen: CHD7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- NOVUNBP1-77393SS
- NOVUNBP1-77393
- Antigen symbol:
- CHD7
- CHD7
- Antigen name:
- Chromodomain helicase DNA binding protein 7
- Chromodomain helicase DNA binding protein 7
- Antigen synonyms:
- HH5|KAL5|ATP-dependent helicase CHD7|chromodomain helicase DNA binding protein 7 isoform CRA_e|IS3|chromodomain-helicase-DNA-binding protein 7
- HH5|KAL5|ATP-dependent helicase CHD7|chromodomain helicase DNA binding protein 7 isoform CRA_e|IS3|chromodomain-helicase-DNA-binding protein 7
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 55636
- 55636
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together