Anti-TP53BP1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: TP53BP1 (tumor protein p53 binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
- Catalog No:
- NOVUNB100-904SS
- NOVUNB100-904
- Antigen symbol:
- 53BP1
- 53BP1
- Antigen name:
- Tumor suppressor p53-binding protein 1
- Tumor suppressor p53-binding protein 1
- Antigen synonyms:
- FLJ41424|TP53BP1|MGC138366|53BP1|p53BP1|p202
- FLJ41424|TP53BP1|MGC138366|53BP1|p53BP1|p202
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 7158
- 7158
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together