Anti-GFP Rabbit Polyclonal Antibody
NOVUNB100-56402
- Antibody type:Tag
- Antigen name:Green Fluorescence Protein
- Antigen symbol:GFP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Multiple
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Tag
Antigen: GFP (green fluorescent protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Pan