Anti-EGR2 Rabbit Polyclonal Antibody
Catalog # BSBTPA2178
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Early growth response protein 2
- Antigen symbol:EGR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:KROX20|Drosophila|homolog (early growth response-2)|KROX-20|Drosophila) |early growth response 2 (Krox-20 homolog|DKFZp686J1957|CMT4E|Krox-20 homolog|OTTHUMP00000062133 |CMT1D|EGR2|FLJ14547|early growth response 2 protein
- Amino acid number:153 - 168
- Sequence:TMSQTQPDLDHLYSPP
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human EGR2 (153-168aa TMSQTQPDLDHLYSPP), different from the related rat and mouse sequences by one amino acid
- Purification:Purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for E3 SUMO-protein ligase EGR2(EGR2) detection. Tested with WB in Human, Mouse, Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: EGR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat