Anti-PMP70 Rabbit Polyclonal Antibody
Catalog # BSBTPA2091
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Peroxisomal Membrane Protein 70
- Antigen symbol:PMP70
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:70 kDa peroxisomal membrane protein|Pxmp1|Abc43|Peroxisomal Membrane Protein 70|Abcd3|Pmp70
- Amino acid number:646 - 659
- Sequence:EFKQITEDTVEFGS
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human PMP70 (646-659aa EFKQITEDTVEFGS), different from the related mouse and rat sequences by one amino acid
- Purification:Purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for ATP-binding cassette sub-family D member 3(ABCD3) detection. Tested with WB, IHC-P in Human, Mouse, Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: PMP70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat