Order Entry
Italy
ContactUsLinkComponent
Anti-GJA1 Mouse Monoclonal Antibody [clone: Connexin 43]
Anti-GJA1 Mouse Monoclonal Antibody [clone: Connexin 43]
Catalog # ABCAAB78055-50
Supplier:  Abcam
Anti-GJA1 Mouse Monoclonal Antibody [clone: Connexin 43]
Catalog # ABCAAB78055-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    gap junction Protein, alpha 1, 43kDa
  • Antigen symbol:
    GJA1
  • Clonality:
    Monoclonal
  • Clone:
    Connexin 43
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    50 µG

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: GJA1 (gap junction protein, alpha 1, 43kDa)
Clonality: monoclonal
Clone: Connexin 43
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse