Anti-ADAMTS4 Rabbit Polyclonal Antibody
BIRBORB96641-50
: Biorbyt
- Antibody type:Primary
- Antigen name:ADAM metallopeptidase with thrombospondin type 1 motif, 4
- Antigen symbol:ADAMTS4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic 19 amino acid peptide from internal region of human ADAMTS4
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Immunochemistry: 8 ?g/ml
Type: Primary
Antigen: ADAMTS4 (ADAM metallopeptidase with thrombospondin type 1 motif, 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: