Anti-AEN Rabbit Polyclonal Antibody
BIRBORB73850-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Apoptosis enhancing nuclease
- Antigen symbol:AEN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Concentration:0,9 mg/ml
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: AEN (apoptosis enhancing nuclease)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human