Anti-M13 Bacteriophage Mouse Monoclonal Antibody
BIRBORB108898-100
: Biorbyt
- Antibody type:Primary
- Antigen name:M13 bacteriophage
- Antigen symbol:M13
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with isolated M13 phage coat proteins and mouse SP2/0-Ag14 myeloma cells.
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: M13 Bacteriophage
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: