Anti-TIAL1/TIA Goat Polyclonal Antibody
BIRBORB20603-100
: Biorbyt
- Antibody type:Primary
- Antigen symbol:TIAL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:TIA
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide (Internal Region)
- Purification:purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: TIAL1/TIA
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: