Anti-NFAM1 (NFAT activating Protein with ITAM motif 1) Mouse Polyclonal Antibody
Catalog # ABNOH00150372-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:NFAT activating Protein with ITAM motif 1
- Antigen symbol:NFAM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:150372
- Antigen synonyms:bK126B4.4|CNAIP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: NFAM1 (NFAT activating Protein with ITAM motif 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human