Anti-CANT1 Mouse Monoclonal Antibody [clone: 1A1]
Catalog # ABNOH00124583-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Calcium Activated Nucleotidase 1
- Antigen symbol:CANT1
- Clonality:Monoclonal
- Clone:1A1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:124583
- Antigen synonyms:DBQD|SCAN1|SHAPY|SCAN-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CANT1
Clonality: Monoclonal
Clone: 1A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human