Anti-ERI1 Mouse Polyclonal Antibody
ABNOH00090459-B01
: Abnova
- Antibody type:Primary
- Antigen name:Exoribonuclease 1
- Antigen symbol:ERI1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:90459
- Antigen synonyms:protei|enhanced RNAi three prime mRNA exonuclease homolog 1|Eri-1 homolog|3'-5' exoribonuclease 1|histone mRNA 3'-exonuclease 1|3'HEXO|histone mRNA 3'-end-specific exoribonuclease|histone mRNA 3' end-specific exonuclease|HEXO|THEX1|3' exoribonuclease|3'-5' exonuclease ERI1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: ERI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human