Anti-PEAK1 Mouse Monoclonal Antibody [clone: 3E7]
Catalog # ABNOH00079834-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Pseudopodium-enriched Atypical Kinase 1
- Antigen symbol:PEAK1
- Clonality:Monoclonal
- Clone:3E7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:79834
- Antigen synonyms:FLJ34483 |FLJ21140|KIAA2002|EC 2.7.10.2|SGK269
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PEAK1
Clonality: Monoclonal
Clone: 3E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human