Anti-FYCO1 Mouse Polyclonal Antibody
ABNOH00079443-B01P
: Abnova
- Antibody type:Primary
- Antigen name:FYVE and coiled-coil domain containing 1
- Antigen symbol:FYCO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:79443
- Antigen synonyms:CTRCT18|CATC2|FYVE and coiled-coil domain-containing protein 1|RUFY3|ZFYVE7|Zinc finger FYVE domain-containing protein 7|FYCO1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: FYCO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human