Order Entry
Italy
ContactUsLinkComponent
 
Anti-ACBD3 Rabbit Polyclonal Antibody
  ABNOH00064746-D01P
 :  Abnova
undefined
Anti-ACBD3 Rabbit Polyclonal Antibody
  ABNOH00064746-D01P
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Acyl-CoA Binding Domain Containing 3
  • Antigen symbol:
    ACBD3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    64746
  • Antigen synonyms:
    GOCAP 1|Peripherial benzodiazepine receptor associated protein|PAP 7|ACBD 3|Golgi complex associated protein 1 60kDa|GCP60|Golgi phosphoprotein 1|GCP 60|Golgi complex-associated protein 1|Acyl Coenzyme A binding domain containing 3|GOLPH 1|GOCAP1|Acyl CoA binding domain containing protein 3|Peripheral benzodiazepine receptor associated protein PAP7|PKA RIalpha associated protein.|PBR associated protein|ACBD3|PAP7|Golgi complex associated protein 1|GCP60_HUMAN|PBR and PKA associated protein 7|Golgi resident protein GCP60|PBR- and PKA-associated protein 7|Acyl-CoA-binding domain-containing protein 3|Peripheral benzodiazepine receptor-associated protein PAP7|GOLPH1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: ACBD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human