Anti-JPH1 Mouse Monoclonal Antibody [clone: 2E6]
Codice catalogo ABNOH00056704-M04
Nome del fornitore: Abnova
Specifiche
- Antibody type:Primary
- Antigen name:Junctophilin 1
- Antigen symbol:JPH1
- Clonality:Monoclonal
- Clone:2E6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:56704
- Antigen synonyms:JPH1|Junctophilin type 1|JP1|Junctophilin-1|JP-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifiche
Informazioni su questo articolo
Type: Primary
Antigen: JPH1
Clonality: Monoclonal
Clone: 2E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human