Anti-TRIM39 Mouse Polyclonal Antibody
Catalog # ABNOH00056658-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Tripartite Motif Containing 39
- Antigen symbol:TRIM39
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:56658
- Antigen synonyms:TRIM39B|TFP|RNF23
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: TRIM39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human