Anti-NPLOC4 (nuclear Protein localization 4 homolog (S. cerevisiae)) Rabbit Polyclonal Antibody
Catalog # ABNOH00055666-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:NPL4 homolog, ubiquitin recognition factor
- Antigen symbol:NPLOC4 (nuclear Protein localization 4 homolog (S. cerevisiae))
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:55666
- Antigen synonyms:NPL4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: NPLOC4 (nuclear Protein localization 4 homolog (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human