Anti-STRADB Mouse Monoclonal Antibody [clone: 3E11]
Catalog # ABNOH00055437-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:STE20-Related Kinase Adapter Beta
- Antigen symbol:STRADB
- Clonality:Monoclonal
- Clone:3E11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:55437
- Antigen synonyms:Als2cr2|AA792893|PRO1038|ILPIPA|D1Ucla2|ILPIP|B830008M19|Syradb|Papk
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: STRADB
Clonality: Monoclonal
Clone: 3E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human