Anti-MED18 Rabbit Polyclonal Antibody
ABNOH00054797-D01
: Abnova
- Antibody type:Primary
- Antigen name:Mediator Complex Subunit 18
- Antigen symbol:MED18
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:54797
- Antigen synonyms:p28b
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: MED18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human