Anti-FBXO40 Mouse Monoclonal Antibody [clone: 2E5]
ABNOH00051725-M01
: Abnova
- Antibody type:Primary
- Antigen name:F-box protein 40
- Antigen symbol:FBXO40
- Clonality:Monoclonal
- Clone:2E5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:51725
- Antigen synonyms:FBX40
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: FBXO40
Clonality: Monoclonal
Clone: 2E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human