Order Entry
Italy
ContactUsLinkComponent
Anti-ANAPC11 Mouse Monoclonal Antibody [clone: 1B4-1A4]
Anti-ANAPC11 Mouse Monoclonal Antibody [clone: 1B4-1A4]
Catalog # ABNOH00051529-M01
Supplier:  Abnova
undefined
Anti-ANAPC11 Mouse Monoclonal Antibody [clone: 1B4-1A4]
Catalog # ABNOH00051529-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Anaphase Promoting Complex Subunit 11
  • Antigen symbol:
    ANAPC11
  • Clonality:
    Monoclonal
  • Clone:
    1B4-1A4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    51529
  • Antigen synonyms:
    Apc11p|HSPC214|APC11
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: ANAPC11
Clonality: Monoclonal
Clone: 1B4-1A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human