Anti-ETV7 Mouse Polyclonal Antibody
ABNOH00051513-B01P
: Abnova
- Antibody type:Primary
- Antigen name:ETS variant 7
- Antigen symbol:ETV7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:51513
- Antigen synonyms:TELB|TEL-2|TEL2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: ETV7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human