Anti-UFC1 Mouse Polyclonal Antibody
ABNOH00051506-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Ubiquitin-fold modifier conjugating enzyme 1
- Antigen symbol:UFC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:51506
- Antigen synonyms:Ubiquitin-fold modier-conjugating enzyme 1|UFC1_HUMAN|CGI-126|Ubiquitin fold modier conjugating enzyme 1|UFC1|HSPC155|Ufm1 conjugating enzyme 1|Ufm1-conjugating enzyme 1.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: UFC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human