Anti-POU2F3 Mouse Polyclonal Antibody
Catalog # ABNOH00025833-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:POU Class 2 Homeobox 3
- Antigen symbol:POU2F3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:25833
- Antigen synonyms:POU transcription factor|MGC126698|Skn-1a|class 2|Epoc-1|OCT11|POU class 2 homeobox 3|PLA-1|transcription factor 3|POU domain|POU2F3|FLJ40063
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: POU2F3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human