Order Entry
Italy
ContactUsLinkComponent
 
Anti-PRDX3 Mouse Monoclonal Antibody [clone: 1E3-B2]
  ABNOH00010935-M01
 :  Abnova
undefined
Anti-PRDX3 Mouse Monoclonal Antibody [clone: 1E3-B2]
  ABNOH00010935-M01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Peroxiredoxin 3
  • Antigen symbol:
    PRDX3
  • Clonality:
    Monoclonal
  • Clone:
    1E3-B2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    10935
  • Antigen synonyms:
    AOP1|MGC24293|AOP-1|MGC104387|SP-22|PRO1748|MER5
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: PRDX3
Clonality: Monoclonal
Clone: 1E3-B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human