Anti-MAGED2 Mouse Monoclonal Antibody [clone: 6G10]
Catalog # ABNOH00010916-M04
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Melanoma Antigen Family D2
- Antigen symbol:MAGED2
- Clonality:Monoclonal
- Clone:6G10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:10916
- Antigen synonyms:11B6|HCA10|BCG-1|MAGE-D2|BCG1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: MAGED2
Clonality: Monoclonal
Clone: 6G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human