Anti-GDF11 Mouse Monoclonal Antibody [clone: 1E6]
Catalog # ABNOH00010220-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Growth differentiation factor 11
- Antigen symbol:GDF11
- Clonality:Monoclonal
- Clone:1E6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:10220
- Antigen synonyms:GDF11|Bone morphogenetic protein 11|BMP11|GDF-11|Growth/differentiation factor 11 [Precursor]
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: GDF11
Clonality: Monoclonal
Clone: 1E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human