Anti-EXOG Mouse Monoclonal Antibody [clone: 2F7]
Catalog # ABNOH00009941-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:endo/exonuclease (5'-3'), endonuclease G-like
- Antigen symbol:EXOG
- Clonality:Monoclonal
- Clone:2F7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:9941
- Antigen synonyms:ENGLA|ENDOGL1|ENGL-b|ENGLB|ENGL|ENGL-a|ENDOGL2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: EXOG
Clonality: Monoclonal
Clone: 2F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat