Anti-MFN2 Mouse Monoclonal Antibody [clone: 6A8]
Catalog # ABNOH00009927-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Mitofusin 2
- Antigen symbol:MFN2
- Clonality:Monoclonal
- Clone:6A8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:9927
- Antigen synonyms:HSG|CMT2A|Transmembrane GTPase MFN2|CMT2A2|MARF|Mitofusin-2|MFN2|KIAA0214|CPRP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: MFN2
Clonality: Monoclonal
Clone: 6A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat