Anti-CD83 Mouse Polyclonal Antibody
ABNOH00009308-B01P
: Abnova
- Antibody type:Primary
- Antigen name:CD83 molecule
- Antigen symbol:CD83
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:9308
- Antigen synonyms:Cell-Surface Glycoprotein|B-Cell Activation Protein|BL11|HB15
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: CD83
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human