Anti-ACOX2 Mouse Polyclonal Antibody
ABNOH00008309-A01
: Abnova
- Antibody type:Primary
- Antigen name:Acyl-CoA Oxidase 2, branched chain
- Antigen symbol:ACOX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:8309
- Antigen synonyms:Trihydroxycoprostanoyl-CoA oxidase|ACOX2|THCA-CoA oxidase|BRCACOX|branched chain |BRCOX|THCCox |acyl-Coenzyme A oxidase 2|BCOX|Peroxisomal branched chain acyl-CoA oxidase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: ACOX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human