Anti-ZIC2 (Zic Family member 2) Mouse Monoclonal Antibody [clone: 3C12]
ABNOH00007546-M01
: Abnova
- Antibody type:Primary
- Antigen name:Zic family member 2
- Antigen symbol:ZIC2 (Zic Family member 2)
- Clonality:Monoclonal
- Clone:3C12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:7546
- Antigen synonyms:HPE5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ZIC2 (Zic Family member 2)
Clonality: Monoclonal
Clone: 3C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human