Anti-VIPR1 Mouse Polyclonal Antibody
ABNOH00007433-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Vasoactive intestinal peptide receptor 1
- Antigen symbol:VIPR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:7433
- Antigen synonyms:VIP-R-1|II|PACAP-R-2|PACAP-R2|VPAC1|VPAC1R|VPCAP1R|HVR1|VAPC1|V1RG|RDC1|VIRG|VIPR
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: VIPR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human