Order Entry
Italy
ContactUsLinkComponent
 
Anti-TGFA Mouse Polyclonal Antibody
  ABNOH00007039-A01
 :  Abnova
undefined
Anti-TGFA Mouse Polyclonal Antibody
  ABNOH00007039-A01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Transforming Growth Factor, Alpha
  • Antigen symbol:
    TGFA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    7039
  • Antigen synonyms:
    transforming growth factor alpha|TGF-alpha|TGFalpha|ETGF|TGF-a|TGF type 1|TGF alpha|EGF-like TGF
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

 

 

Type: Primary
Antigen: TGFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human