Anti-RORC Mouse Monoclonal Antibody [clone: 1G7]
Catalog # ABNOH00006097-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Retinoid related orphan receptor gamma
- Antigen symbol:RORC
- Clonality:Monoclonal
- Clone:1G7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6097
- Antigen synonyms:RZRG|NR1F3|RORG|RZR-GAMMA|TOR
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: RORC
Clonality: Monoclonal
Clone: 1G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human