Order Entry
Italy
ContactUsLinkComponent
 
Anti-MAP2K3 Rabbit Polyclonal Antibody
  ABNOH00005606-D01P
 :  Abnova
undefined
Anti-MAP2K3 Rabbit Polyclonal Antibody
  ABNOH00005606-D01P
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase Kinase 3
  • Antigen symbol:
    MAP2K3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    5606
  • Antigen synonyms:
    dual specificity mitogen-activated protein kinase kinase 3|SAPKK-2|MEK 3|MAP kinase kinase 3|MAPKK3|SAPK kinase 2|SAPKK2|PRKMK3|stress-activated protein kinase kinase 2|MKK3|MAPKK 3|MAPK/ERK kinase 3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: MAP2K3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human