Anti-IFIT3 Mouse Polyclonal Antibody
50 µg
Catalog # ABNOH00003437-B01P
Supplier: Abnova
Jump to:
Product Details & Documents
Type: Primary
Antigen: IFIT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Size:50 µg
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Storage temperature:Cool
- Host:Mouse
- Cross adsorption:No
- Shipping temperature:Ice
- Antigen name:Interferon-induced protein with tetratricopeptide repeats 3
- Cat. No.:ABNOH00003437-B01P
- Reactivity:Human
- Gene ID:3437
- Antigen synonyms:IFIT-3|IFIT4|CIG-49|IFI60|RIG-G|Interferon-induced protein with tetratricopeptide repeats 3|IRG2|IFIT3 |P60|ISG60|GARG-49|CIG49
- Conjugation:Unconjugated
- Antigen symbol:IFIT3



