Anti-FUT2 Rabbit Polyclonal Antibody
ABNOH00002524-D01
: Abnova
- Antibody type:Primary
- Antigen name:Fucosyltransferase 2
- Antigen symbol:FUT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2524
- Antigen synonyms:fucosyltransferase 2|alpha|SEC2|Se2|12FT2|galactoside 2-alpha-L-fucosyltransferase 2|GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 2|sej|B12QTL1|12 fucosyltransferase|12FT 2|secretor status included|SE
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: FUT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human