Anti-STOM Mouse Polyclonal Antibody
ABNOH00002040-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Stomatin
- Antigen symbol:STOM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2040
- Antigen synonyms:STOM|EPB7|protein 7.2b|EPB72|erythrocyte membrane protein band 7.2|erythrocyte band 7 integral membrane protein|BND7
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: STOM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human