Anti-CYP4B1 Rabbit Polyclonal Antibody
ABNOH00001580-D01P
: Abnova
- Antibody type:Primary
- Antigen name:Cytochrome P450, Family 4, Subfamily B, Polypeptide 1
- Antigen symbol:CYP4B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:1580
- Antigen synonyms:Cytochrome P450 4B1|CYPIVB1|Cytochrome P450-HP|CYP4B1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: CYP4B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse