Order Entry
Italy
ContactUsLinkComponent
Anti-CBR3 Rabbit Polyclonal Antibody
Anti-CBR3 Rabbit Polyclonal Antibody
  ABNOH00000874-D01
 :  Abnova
undefined
Anti-CBR3 Rabbit Polyclonal Antibody
  ABNOH00000874-D01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Carbonyl Reductase 3
  • Antigen symbol:
    CBR3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    874
  • Antigen synonyms:
    hCBR3|SDR21C2|HEL-S-25
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: CBR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human