Order Entry
Italy
ContactUsLinkComponent
 
Anti-NKX3-2 Mouse Polyclonal Antibody
Catalog # ABNOH00000579-A01
Supplier:  Abnova
undefined
Anti-NKX3-2 Mouse Polyclonal Antibody
Catalog # ABNOH00000579-A01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    NK3 homeobox 2
  • Antigen symbol:
    NKX3-2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    579
  • Antigen synonyms:
    BAPX1|NKX3.2|Bagpipe homeobox protein homolog 1|NKX3B|SMMD
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: NKX3-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human