Anti-SERPINB3 Mouse Monoclonal Antibody [clone: OTI1A12]
Catalog # ORIGTA506878
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Serpin peptidase inhibitor, clade B (ovalbumin), member 3
- Antigen symbol:SERPINB3
- Clonality:Monoclonal
- Clone:OTI1A12
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:44.4 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150
Type: Primary
Antigen: SERPINB3 (serpin peptidase inhibitor, clade B (ovalbumin), member 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human