Anti-NRXN1 Goat Polyclonal Antibody
ORIGTA302549
: OriGene
- Antibody type:Primary
- Antigen name:Neurexin 1
- Antigen symbol:NRXN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Form:Liquid
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Molecular weight:50.49 kDa
- Sequence:C-KEKQPSSAKSSNKN
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Pk:100 µG
Recommended Dilutions: ELISA: 1:32000; Western Blot: 005-01µg/ml
Type: Primary
Antigen: NRXN1 (neurexin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat