Order Entry
Italy
ContactUsLinkComponent
Anti-KLRB1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: PK136]
Anti-KLRB1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: PK136]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-KLRB1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: PK136]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 35-5941-U100
  • 35-5941-U500
  • 35-5941-U025
  • Antigen symbol:
  • KLRB1C
  • KLRB1C
  • KLRB1C
  • Antigen name:
  • Killer Cell Lectin-Like Receptor Subfamily B, Member 1
  • Killer Cell Lectin-Like Receptor Subfamily B, Member 1
  • Killer Cell Lectin-Like Receptor Subfamily B, Member 1
  • Antigen synonyms:
  • NK-RP1|ly-55c|Nk1|AI462337|NKR-P1.9|Ly-59|Nk-1.2|NK1.1|Ly55c|Ly59|Nk-1|Nkrp1c|NKRP140|CD161|Nk1.2|NKRP1
  • NK-RP1|ly-55c|Nk1|AI462337|NKR-P1.9|Ly-59|Nk-1.2|NK1.1|Ly55c|Ly59|Nk-1|Nkrp1c|NKRP140|CD161|Nk1.2|NKRP1
  • NK-RP1|ly-55c|Nk1|AI462337|NKR-P1.9|Ly-59|Nk-1.2|NK1.1|Ly55c|Ly59|Nk-1|Nkrp1c|NKRP140|CD161|Nk1.2|NKRP1
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • PK136
  • PK136
  • PK136
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 17059
  • 17059
  • 17059
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together