Anti-S100A10 Rabbit Polyclonal Antibody
ABGEAP9090C
: Abgent
- Antibody type:Primary
- Antigen name:S100 Calcium Binding Protein A10
- Antigen symbol:S100A10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Center
- Gene ID:245650
- Antigen synonyms:42C|GP11|CLP11|ANX2L|CAL1L|Ca[1]|p10|P11|ANX2LG
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:ARGEIPEKPDSNQC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,46mg/ml
- Shipping temperature:4 °C
- Immunogen:This Mouse Gucy2f antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 317-344 amino acids from the Central region of mouse Gucy2f.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: S100A10 (S100 calcium binding protein A10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human